Gene Transcripts

Gene Symbol
RASGRF1
Entrez Gene ID
5923
Gene Name
Ras protein specific guanine nucleotide releasing factor 1
Gene Aliases
CDC25, CDC25L, GNRP, GRF1, GRF55, H-GRF55, PP13187, ras-GRF1
Chromosome Location
Chr. 15: 78959947 - 79090873 on Build GRCh38
UniGene ID
Hs.459035
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145648.1 NP_001139120.1 22 3580
NM_002891.4 NP_002882.3 23 3628
NM_153815.2 NP_722522.1 9 2074
XM_011521866.2 XP_011520168.1 23 3528
XM_011521867.2 XP_011520169.1 23 3528
XM_017022455.1 XP_016877944.1 23 3519
XM_017022456.1 XP_016877945.1 22 3489
XM_017022457.1 XP_016877946.1 22 3489
XM_017022458.1 XP_016877947.1 22 3480
XM_017022459.1 XP_016877948.1 22 3489
GenBank mRNA AK226101.1 - 21 2864
AK301205.1 - 11 1677
AK309047.1 - 21 3011
BC040275.1 - 22 3471 5192914
L26584.1 - 23 3259
M91815.1 - 2 121
S62035.1 - 9 2083

View Full Product Details