Gene Transcripts

Gene Symbol
Chrna10
Entrez Gene ID
504186
Gene Name
cholinergic receptor, nicotinic, alpha polypeptide 10
Gene Aliases
Chromosome Location
Chr. 7: 102111262 - 102116828 on Build GRCm38
UniGene ID
Mm.334074
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081424.1 NP_001074893.1 2 500
GenBank mRNA AK033068.1 - 2 501
BC147245.1 - 2 215 9056293
BC147246.1 - 2 215 9056310
BC151033.1 - 2 215 9087946
BC151035.1 - 2 215 9087948

View Full Product Details