Gene Transcripts
- Gene Symbol
- Baiap3
- Entrez Gene ID
- 545192
- Gene Name
- BAI1-associated protein 3
- Gene Aliases
- Bap3, Gm937
- Chromosome Location
- Chr. 17: 25242659 - 25256364 on Build GRCm38
- UniGene ID
- Mm.328859
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK122358.1 |
- |
13 |
1216 |
|
BC158116.1 |
- |
13 |
1314 |
9088189 |
View Full Product Details