Gene Transcripts

Gene Symbol
Cyp3a57
Entrez Gene ID
622127
Gene Name
cytochrome P450, family 3, subfamily a, polypeptide 57
Gene Aliases
EG622127
Chromosome Location
Chr. 5: 145345145 - 145390926 on Build GRCm38
UniGene ID
Mm.359379
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100180.1 NP_001093650.1 13 1526

View Full Product Details