Gene Transcripts
- Gene Symbol
- Trim63
- Entrez Gene ID
- 433766
- Gene Name
- tripartite motif-containing 63
- Gene Aliases
- MuRF1, RF1, Rnf28
- Chromosome Location
- Chr. 4: 134315088 - 134329629 on Build GRCm38
- UniGene ID
- Mm.331961
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC147663.1 |
- |
7 |
1155 |
|
BC147666.1 |
- |
7 |
1155 |
9087455 |
View Full Product Details