Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003013.2 | NP_003004.1 | 1 | 485 | |
GenBank mRNA | AF017986.1 | - | 1 | 508 | |
AF311912.1 | - | 1 | 481 | ||
AK075372.1 | - | 1 | 478 | ||
AK093922.1 | - | 2 | 453 | ||
AY359001.1 | - | 1 | 287 | ||
BC008666.1 | - | 1 | 465 | 3874262 | |
BM906663.1 | - | 544 | |||
BM924815.1 | - | 1 | 500 | ||
CN431016.1 | - | 1 | 494 |