Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012522.2 | NP_036654.2 | 9 | 1022 | |
GenBank mRNA | D01098.1 | - | 9 | 1023 | |
FQ211441.1 | - | 9 | 1081 | ||
M88344.1 | - | 9 | 1171 | ||
M88345.1 | - | 6 | 1435 | ||
M88346.1 | - | 8 | 917 | ||
M88347.1 | - | 8 | 917 |