Gene Transcripts

Gene Symbol
Cyp21a1
Entrez Gene ID
24298
Gene Name
cytochrome P450, family 21, subfamily a, polypeptide 1
Gene Aliases
21-OH, Cyp21
Chromosome Location
Chr. 20: 4486213 - 4489358 on Build Rnor_6.0
UniGene ID
Rn.161726
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057101.2 NP_476442.2 8 962
XM_006255907.2 XP_006255969.1 8 1036
GenBank mRNA U56853.1 - 8 963
U56854.1 - 7 1024

View Full Product Details