Gene Transcripts

Gene Symbol
SHBG
Entrez Gene ID
6462
Gene Name
sex hormone binding globulin
Gene Aliases
ABP, SBP, TEBG
Chromosome Location
Chr. 17: 7614064 - 7633383 on Build GRCh38
UniGene ID
Hs.632235
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040.4 NP_001031.2 3 398
NM_001146279.2 NP_001139751.1 3 398
NM_001146280.2 NP_001139752.1 3 398
NM_001146281.2 NP_001139753.1 3 398
NM_001289113.1 NP_001276042.1 3 391
NM_001289114.1 NP_001276043.1 3 254
NM_001289115.1 NP_001276044.1 3 391
NM_001289116.1 NP_001276045.1 3 398
GenBank mRNA AK302603.1 - 3 391
BC069597.1 - 3 316
BC101785.1 - 3 350 8069291
BC112186.1 - 3 350 8327654
CD013955.1 - 3 340
CD013957.1 - 3 340
CD013958.1 - 3 340
EU352661.1 - 3 238
EU352662.1 - 3 238
EU352663.1 - 3 238
EU352664.1 - 3 238
EU352665.1 - 3 259
EU352666.1 - 3 259
EU352667.1 - 3 259
EU352668.1 - 3 259
EU352669.1 - 3 259
EU352670.1 - 3 254
EU352671.1 - 3 254
EU352672.1 - 3 254
EU352673.1 - 3 254
EU352674.1 - 3 254
EU352675.1 - 3 254
EU352676.1 - 3 254
X05403.1 - 3 256
X05885.1 - 2 183
X16351.1 - 3 304

View Full Product Details