Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199692.1 | NP_001186621.1 | 18 | 3885 | |
NM_001199693.1 | NP_001186622.1 | 17 | 2911 | ||
NM_001199694.1 | NP_001186623.1 | 17 | 3332 | ||
NM_003040.3 | NP_003031.3 | 18 | 3101 | ||
GenBank mRNA | AB209158.1 | - | 18 | 3885 | |
AK295767.1 | - | 17 | 2911 | ||
AK312699.1 | - | 18 | 3080 | ||
BC004893.2 | - | 18 | 2738 | 3507709 | |
BC009386.2 | - | 17 | 3332 | 4127724 | |
BC009434.1 | - | 17 | 3332 | 3501642 | |
BC010069.2 | - | 18 | 2738 | 4125444 | |
BC028601.2 | - | 17 | 2933 | 4814066 | |
U62531.1 | - | 18 | 3019 | ||
X03918.1 | - | 11 | 1717 | ||
X62137.1 | - | 18 | 2921 |