Gene Transcripts

Gene Symbol
Cpt1b
Entrez Gene ID
25756
Gene Name
carnitine palmitoyltransferase 1b, muscle
Gene Aliases
CPT-IB, M-CPTI
Chromosome Location
Chr. 7: 130395211 - 130404718 on Build Rnor_6.0
UniGene ID
Rn.6028
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013200.1 NP_037332.1 16 1953
XM_006242180.2 XP_006242242.1 16 2409
GenBank mRNA BC085761.1 - 16 1965
D43623.1 - 15 1940

View Full Product Details