Gene Transcripts

Gene Symbol
Spr
Entrez Gene ID
29270
Gene Name
sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase)
Gene Aliases
Chromosome Location
Chr. 4: 116912343 - 116916073 on Build Rnor_6.0
UniGene ID
Rn.6658
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019181.1 NP_062054.1 3 658
GenBank mRNA BC166809.1 - 3 658
DY314075.1 - 3 657
FQ226380.1 - 3 495
FQ227075.1 - 3 508
M36410.1 - 3 634

Gene Symbol
RGD1560795
Entrez Gene ID
297402
Gene Name
similar to Sepiapterin reductase (SPR)
Gene Aliases
Chromosome Location
Chr. 4: 116933277 - 116936466 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006236796.2 XP_006236858.1 2 446

View Full Product Details