Gene Transcripts

Gene Symbol
SNRPC
Entrez Gene ID
6631
Gene Name
small nuclear ribonucleoprotein polypeptide C
Gene Aliases
U1C, Yhc1
Chromosome Location
Chr. 6: 34757094 - 34773857 on Build GRCh38
UniGene ID
Hs.1063
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003093.2 NP_003084.1 3, 4 613
NR_029472.1 2, 3 570
GenBank mRNA AK223098.1 - 3, 4 187
AK311955.1 - 3, 4 193
BC121082.1 - 3, 4 182 40119776
BC121083.1 - 3, 4 182 40119777
BU153661.1 - 2, 3 178
BU508721.1 - 2, 3 139
CD109745.1 - 3, 4 630
M18465.1 - 1, 2 91
X12517.1 - 3, 4 178

View Full Product Details