Gene Transcripts

Gene Symbol
STK10
Entrez Gene ID
6793
Gene Name
serine/threonine kinase 10
Gene Aliases
LOK, PRO2729
Chromosome Location
Chr. 5: 172042070 - 172188342 on Build GRCh38
UniGene ID
Hs.744005
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005990.3 NP_005981.3 7 1123
XM_017009788.1 XP_016865277.1 6 677
GenBank mRNA AB015718.1 - 7 873
AK307943.1 - 7 657
AK313350.1 - 8 878
BC070077.1 - 7 1037 5267449
DB068209.1 - 2 140

View Full Product Details