Gene Transcripts

Gene Symbol
Kcnma1
Entrez Gene ID
83731
Gene Name
potassium channel, calcium activated large conductance subfamily M alpha, member 1
Gene Aliases
BKCa, KCNMA1b, KCNMA1c, Kcnma, Slo
Chromosome Location
Chr. 15: 344204 - 1048849 on Build Rnor_6.0
UniGene ID
Rn.30616
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031828.1 NP_114016.1 11 1906
GenBank mRNA AB248960.1 - 4 322
AF083341.1 - 11 1342
AF135265.1 - 11 1760
AY330290.1 - 11 1231
AY330291.1 - 11 1231
AY330293.2 - 11 1231
AY330294.1 - 11 1231
AY344964.1 - 11 1231
AY344965.2 - 10 1643
U40603.1 - 11 1906
U55995.1 - 11 1384
U93052.1 - 11 1332

View Full Product Details