Gene Transcripts

Gene Symbol
Ptms
Entrez Gene ID
83801
Gene Name
parathymosin
Gene Aliases
ZnBP
Chromosome Location
Chr. 4: 157435371 - 157439507 on Build Rnor_6.0
UniGene ID
Rn.3609
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031975.2 NP_114181.1 2 429
GenBank mRNA BC167753.1 - 2 419
M20616.1 - 1 33
M33025.1 - 1 33
X16481.1 - 2 212

View Full Product Details