Gene Transcripts
- Gene Symbol
- Rps16
- Entrez Gene ID
- 140655
- Gene Name
- ribosomal protein S16
- Gene Aliases
- Chromosome Location
- Chr. 1: 85405499 - 85408444 on Build Rnor_6.0
- UniGene ID
- Rn.29791
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001169146.1 |
NP_001162617.1
|
1, 2 |
113 |
|
XM_006228612.2 |
XP_006228674.1
|
1, 2 |
110 |
|
GenBank mRNA |
BC064030.1 |
- |
|
87 |
|
BC084715.1 |
- |
1, 2 |
82 |
|
BC159438.1 |
- |
1, 2 |
84 |
|
FQ211832.1 |
- |
1, 2 |
94 |
|
FQ221089.1 |
- |
1, 2 |
101 |
|
FQ221637.1 |
- |
1, 2 |
95 |
|
FQ221660.1 |
- |
1, 2 |
94 |
|
FQ221692.1 |
- |
1, 2 |
92 |
|
FQ222913.1 |
- |
1, 2 |
94 |
|
FQ223100.1 |
- |
1, 2 |
95 |
|
FQ223384.1 |
- |
1, 2 |
94 |
|
FQ223396.1 |
- |
1, 2 |
95 |
|
FQ224101.1 |
- |
1, 2 |
94 |
|
FQ228433.1 |
- |
1, 2 |
95 |
|
FQ229060.1 |
- |
1, 2 |
47 |
|
X17665.1 |
- |
1, 2 |
93 |
|
View Full Product Details