Gene Transcripts

Gene Symbol
Vom2r75
Entrez Gene ID
286986
Gene Name
vomeronasal 2 receptor, 75
Gene Aliases
Chromosome Location
Chr. 18: 11936 - 38488 on Build Rnor_6.0
UniGene ID
Rn.10813
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173320.1 NP_775442.1 1 176
XM_006254377.2 XP_006254439.1 1 290
GenBank mRNA AF016185.1 - 1 176

View Full Product Details