Gene Transcripts
- Gene Symbol
- Vom2r75
- Entrez Gene ID
- 286986
- Gene Name
- vomeronasal 2 receptor, 75
- Gene Aliases
- Chromosome Location
- Chr. 18: 11936 - 38488 on Build Rnor_6.0
- UniGene ID
- Rn.10813
- Species
- Rattus norvegicus
- Gene Symbol
- LOC286980
- Entrez Gene ID
- 286980
- Gene Name
- putative pheromone receptor (Go-VN13B)
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Rn.10813
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF016186.1 |
- |
4 |
1595 |
|
View Full Product Details