Gene Transcripts

Gene Symbol
Armc7
Entrez Gene ID
287827
Gene Name
armadillo repeat containing 7
Gene Aliases
RGD1310043
Chromosome Location
Chr. 10: 104036250 - 104052852 on Build Rnor_6.0
UniGene ID
Rn.198899
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127522.1 NP_001120994.1 2 434
XM_006247711.2 XP_006247773.1 2 551
GenBank mRNA BC166454.1 - 2 408

View Full Product Details