Gene Transcripts

Gene Symbol
TP53BP2
Entrez Gene ID
7159
Gene Name
tumor protein p53 binding protein 2
Gene Aliases
53BP2, ASPP2, BBP, P53BP2, PPP1R13A
Chromosome Location
Chr. 1: 223779893 - 223845981 on Build GRCh38
UniGene ID
Hs.523968
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031685.2 NP_001026855.2 12 2206
NM_005426.2 NP_005417.1 13 2338
XM_011544268.1 XP_011542570.1 11 2067
XM_011544269.2 XP_011542571.1 9 1725
XM_017002220.1 XP_016857709.1 13 2347
GenBank mRNA AJ318888.2 - 12 2151
AK294432.1 - 12 2058
AK294964.1 - 5 1226
AK295378.1 - 6 930
AK316016.1 - 5 1128
AK316103.1 - 5 1106
BC040247.1 - 10 1682 4454266
BC046150.1 - 12 2149
BC058918.1 - 12 2164 4537206
U09582.1 - 1 101
U58334.1 - 13 2283

View Full Product Details