Gene Transcripts
- Gene Symbol
- TPTE
- Entrez Gene ID
- 7179
- Gene Name
- transmembrane phosphatase with tensin homology
- Gene Aliases
- CT44, PTEN2
- Chromosome Location
- Chr. 21: 10521514 - 10606270 on Build GRCh38
- UniGene ID
- Hs.122986
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290224.1 |
NP_001277153.1
|
16, 17 |
1469 |
|
NM_199259.3 |
NP_954868.1
|
20, 21 |
1691 |
|
NM_199260.3 |
NP_954869.1
|
19, 20 |
1631 |
|
NM_199261.3 |
NP_954870.2
|
21, 22 |
1745 |
|
GenBank mRNA |
AF007118.1 |
- |
21, 22 |
1693 |
|
AF495908.1 |
- |
20, 21 |
1639 |
|
AK308254.1 |
- |
20, 21 |
1588 |
|
AK314285.1 |
- |
21, 22 |
1644 |
|
AL833345.1 |
- |
21, 22 |
1652 |
|
AY219886.1 |
- |
20, 21 |
1639 |
|
AY219887.1 |
- |
19, 20 |
1579 |
|
AY219888.1 |
- |
16, 17 |
1417 |
|
BC028719.1 |
- |
20, 21 |
1674 |
5269598 |
View Full Product Details