Gene Transcripts

Gene Symbol
Orc6
Entrez Gene ID
291927
Gene Name
origin recognition complex, subunit 6
Gene Aliases
Orc6l
Chromosome Location
Chr. 19: 27457536 - 27464804 on Build Rnor_6.0
UniGene ID
Rn.41300
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033690.1 NP_001028862.1 7 1008
XM_006255357.1 XP_006255419.1 6 598
GenBank mRNA BC101871.1 - 7 1008

View Full Product Details