Gene Transcripts

Gene Symbol
Rbm8a
Entrez Gene ID
295284
Gene Name
RNA binding motif protein 8A
Gene Aliases
Rbm8
Chromosome Location
Chr. 2: 198755261 - 198758028 on Build Rnor_6.0
UniGene ID
Rn.37716
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271138.1 NP_001258067.1 5 452
XM_006232956.2 XP_006233018.1 5 450
GenBank mRNA DQ359102.1 - 5 404
FQ210627.1 - 5 425

View Full Product Details