Gene Transcripts

Gene Symbol
ZNF12
Entrez Gene ID
7559
Gene Name
zinc finger protein 12
Gene Aliases
GIOT-3, HZF11, KOX3, ZNF325
Chromosome Location
Chr. 7: 6688433 - 6706935 on Build GRCh38
UniGene ID
Hs.431471
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006956.2 NP_008887.2 6 1859
NM_016265.3 NP_057349.2 5 1973
GenBank mRNA AF505656.1 - 5 1638
AK022691.1 - 1 823
AK293925.1 - 6 1619
AY500354.1 - 2 1275
BC051892.1 - 6 1847
BC137068.1 - 4 1465 9021065
BC137069.1 - 4 1465 9021066
BC144465.1 - 5 1518
BC154414.1 - 5 1961 40148925
BX640749.1 - 5 1384

View Full Product Details