Gene Transcripts

Gene Symbol
Pus7
Entrez Gene ID
296751
Gene Name
pseudouridylate synthase 7
Gene Aliases
RGD1307054
Chromosome Location
Chr. 4: 7889737 - 7931357 on Build Rnor_6.0
UniGene ID
Rn.230290
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170589.1 NP_001164060.1 14 1757
XM_008762634.1 XP_008760856.1 14 1889

View Full Product Details