Gene Transcripts

Gene Symbol
Eda2r
Entrez Gene ID
296872
Gene Name
ectodysplasin A2 receptor
Gene Aliases
RGD1564025
Chromosome Location
Chr. X: 66560221 - 66602509 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006257051.2 XP_006257113.1 4 472
XM_006257052.2 XP_006257114.1 4 472
XM_008773299.1 XP_008771521.1 4 472

View Full Product Details