Gene Transcripts

Gene Symbol
ZNF33B
Entrez Gene ID
7582
Gene Name
zinc finger protein 33B
Gene Aliases
KOX2, KOX31, ZNF11B
Chromosome Location
Chr. 10: 42574169 - 42638837 on Build GRCh38
UniGene ID
Hs.435774
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001305033.1 NP_001291962.1 4 731
NM_001305035.1 NP_001291964.1 6 861
NM_001305036.1 NP_001291965.1 5 789
NM_001305037.1 NP_001291966.1 5 716
NM_001305038.1 NP_001291967.1 5 808
NM_001305039.1 NP_001291968.1 6 866
NM_001305040.1 NP_001291969.1 7 942
NM_006955.2 NP_008886.1 5 784
GenBank mRNA AJ491697.1 - 5 491
AK304306.1 - 5 415
BC048313.2 - 5 489
BC065181.1 - 6 543
BC107726.1 - 5 482
BG536481.1 - 4 446
CD652356.1 - 5 535
DA230792.1 - 5 521
DB156590.1 - 5 448
DR000119.1 - 7 627
X68684.1 - 2 151

View Full Product Details