Gene Transcripts

Gene Symbol
Snx30
Entrez Gene ID
298033
Gene Name
sorting nexin family member 30
Gene Aliases
RGD1311367
Chromosome Location
Chr. 5: 77052484 - 77157915 on Build Rnor_6.0
UniGene ID
Rn.109496
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106651.1 NP_001100121.1 4 715

View Full Product Details