Gene Transcripts

Gene Symbol
Kcnq4
Entrez Gene ID
298496
Gene Name
potassium channel, voltage-gated KQT-like subfamily Q, member 4
Gene Aliases
Chromosome Location
Chr. 5: 139625783 - 139677300 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764109.1 XP_008762331.1 9 1146
XM_008764111.1 XP_008762333.1 9 871

View Full Product Details