Gene Transcripts

Gene Symbol
Vom2r57
Entrez Gene ID
299683
Gene Name
vomeronasal 2 receptor, 57
Gene Aliases
RGD1562007
Chromosome Location
Chr. 7: 19074068 - 19119476 on Build Rnor_6.0
UniGene ID
Rn.198782
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099474.1 NP_001092944.1 4 1452
XM_008765199.1 XP_008763421.1 5 1461

Gene Symbol
Vom2r56
Entrez Gene ID
503127
Gene Name
vomeronasal 2 receptor, 56
Gene Aliases
RGD1559561
Chromosome Location
Chr. 7: 19543611 - 19559251 on Build Rnor_6.0
UniGene ID
Rn.217634
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099484.1 NP_001092954.1 1452

View Full Product Details