Gene Transcripts

Gene Symbol
Lonrf2
Entrez Gene ID
301361
Gene Name
LON peptidase N-terminal domain and ring finger 2
Gene Aliases
RGD1562137
Chromosome Location
Chr. 9: 45468982 - 45506413 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006244869.2 XP_006244931.1 5 1278

View Full Product Details