Gene Transcripts
- Gene Symbol
- Klhl34
- Entrez Gene ID
- 302703
- Gene Name
- kelch-like family member 34
- Gene Aliases
- RGD1560439
- Chromosome Location
- Chr. X: 39952576 - 39956865 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_228962.8 |
XP_228962.2
|
1 |
1801 |
|
View Full Product Details