Gene Transcripts

Gene Symbol
Kif18b
Entrez Gene ID
303575
Gene Name
kinesin family member 18B
Gene Aliases
RGD1310360
Chromosome Location
Chr. 10: 91008294 - 91027765 on Build Rnor_6.0
UniGene ID
Rn.112739
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039019.1 NP_001034108.1 7, 8 1227
XM_006247505.2 XP_006247567.1 7, 8 1235
XM_008768283.1 XP_008766505.1 7, 8 1236
GenBank mRNA BC099126.1 - 7, 8 1227

View Full Product Details