Gene Transcripts

Gene Symbol
AAAS
Entrez Gene ID
8086
Gene Name
aladin WD repeat nucleoporin
Gene Aliases
AAA, AAASb, ADRACALA, ADRACALIN, ALADIN, GL003
Chromosome Location
Chr. 12: 53307456 - 53321628 on Build GRCh38
UniGene ID
Hs.369144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173466.1 NP_001166937.1 10 1101
NM_015665.5 NP_056480.1 11 1200
XM_011538778.1 XP_011537080.1 11 1132
XM_011538780.1 XP_011537082.1 10 1033
GenBank mRNA AF226048.1 - 11 1119
AJ289857.1 - 11 1127
AK000833.1 - 11 1127
AK094302.1 - 9 1380
AK222509.1 - 11 1127
AK293342.1 - 10 1058
AL110160.1 - 5 411
AY237818.1 - 10 1069
BC000659.2 - 11 1162 3347423
BC018836.2 - 11 1162
BT006912.1 - 11 1037

View Full Product Details