Gene Transcripts

Gene Symbol
Mboat4
Entrez Gene ID
306515
Gene Name
membrane bound O-acyltransferase domain containing 4
Gene Aliases
RGD1566335
Chromosome Location
Chr. 16: 61782797 - 61791091 on Build Rnor_6.0
UniGene ID
Rn.218502
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107317.2 NP_001100787.2 3 1169
GenBank mRNA EU518497.1 - 3 1169

View Full Product Details