Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008766019.1 | XP_008764241.1 | 6 | 1412 | |
XM_008766020.1 | XP_008764242.1 | 6 | 1412 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008771846.1 | XP_008770068.1 | 1 | 779 |