Gene Transcripts

Gene Symbol
Slc12a7
Entrez Gene ID
308069
Gene Name
solute carrier family 12 (potassium/chloride transporter), member 7
Gene Aliases
Kcc4
Chromosome Location
Chr. 1: 32086755 - 32167946 on Build Rnor_6.0
UniGene ID
Rn.64199
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013144.2 NP_001013162.2 4 412
XM_006227789.2 XP_006227851.1 4 537
GenBank mRNA AY429042.1 - 4 290
BC086339.1 - 4 411

View Full Product Details