Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001191623.2 | NP_001178552.2 | 6 | 662 | |
XM_006228797.1 | XP_006228859.1 | 6 | 724 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008759279.1 | XP_008757501.1 | 5 | 728 |