Gene Transcripts

Gene Symbol
OR1E2
Entrez Gene ID
8388
Gene Name
olfactory receptor family 1 subfamily E member 2
Gene Aliases
OR17-135, OR17-136, OR17-93, OR1E4, OR1E7, OST529
Chromosome Location
Chr. 17: 3432870 - 3433841 on Build GRCh38
UniGene ID
Hs.533931
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003554.2 NP_003545.1 1 232
GenBank mRNA BC121105.1 - 1 236
BC121106.1 - 1 236 40120314

View Full Product Details