Gene Transcripts

Gene Symbol
Znf48
Entrez Gene ID
103690203
Gene Name
zinc finger protein 48
Gene Aliases
Chromosome Location
Chr. 1: 198680439 - 198685151 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759934.1 XP_008758156.1 2 872

View Full Product Details