Gene Transcripts

Gene Symbol
Sfrp2
Entrez Gene ID
310552
Gene Name
secreted frizzled-related protein 2
Gene Aliases
Chromosome Location
Chr. 2: 182723732 - 182731277 on Build Rnor_6.0
UniGene ID
Rn.102416
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100700.1 NP_001094170.1 2 754
GenBank mRNA BC161900.1 - 2 754

View Full Product Details