Gene Transcripts

Gene Symbol
Gja10
Entrez Gene ID
313126
Gene Name
gap junction protein, alpha 10
Gene Aliases
Cx-57, Cx57, RGD1309630
Chromosome Location
Chr. 5: 47835066 - 47840953 on Build Rnor_6.0
UniGene ID
Rn.154630
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173508.1 NP_001166979.1 1 1438
XM_008763619.1 XP_008761841.1 2 1817

View Full Product Details