Gene Transcripts

Gene Symbol
Pigo
Entrez Gene ID
313341
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class O
Gene Aliases
Chromosome Location
Chr. 5: 58461055 - 58470693 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006238127.2 XP_006238189.1 7 2600
XM_233141.8 XP_233141.7 7 3143

View Full Product Details