Gene Transcripts

Gene Symbol
Fhl3
Entrez Gene ID
313582
Gene Name
four and a half LIM domains 3
Gene Aliases
Chromosome Location
Chr. 5: 142688970 - 142696330 on Build Rnor_6.0
UniGene ID
Rn.229136
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107979.1 NP_001101449.1 5 717
XM_008764016.1 XP_008762238.1 5 851
XM_008764017.1 XP_008762239.1 5 778

View Full Product Details