Gene Transcripts

Gene Symbol
Sf3a3
Entrez Gene ID
313583
Gene Name
splicing factor 3a, subunit 3
Gene Aliases
LRRGT00093
Chromosome Location
Chr. 5: 142702721 - 142722350 on Build Rnor_6.0
UniGene ID
Rn.16732
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025698.1 NP_001020869.1 7 559
GenBank mRNA BC099183.1 - 7 559

View Full Product Details