Gene Transcripts
- Gene Symbol
- Exph5
- Entrez Gene ID
- 315663
- Gene Name
- exophilin 5
- Gene Aliases
- RGD1560308
- Chromosome Location
- Chr. 8: 57885931 - 57962574 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_236272.6 |
XP_236272.4
|
6 |
4980 |
|
View Full Product Details