Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003730.4 | NP_003721.2 | 5 | 709 | |
XM_017011397.1 | XP_016866886.1 | 6 | 488 | ||
XM_017011398.1 | XP_016866887.1 | 4 | 610 | ||
XM_017011399.1 | XP_016866888.1 | 5 | 670 | ||
GenBank mRNA | AJ419865.1 | - | 5 | 663 | |
AJ419866.1 | - | 5 | 1383 | ||
AK001769.1 | - | 2 | 284 | ||
AK315467.1 | - | 5 | 670 | ||
BC001660.2 | - | 5 | 635 | 2959583 | |
BC001819.2 | - | 5 | 635 | 2959583 | |
BC036282.1 | - | 5 | 696 | ||
BC039713.1 | - | 5 | 638 | 5225327 | |
BC051912.1 | - | 5 | 648 | 5227463 | |
U85625.2 | - | 5 | 666 |