Gene Transcripts

Gene Symbol
RNASET2
Entrez Gene ID
8635
Gene Name
ribonuclease T2
Gene Aliases
RNASE6PL, bA514O12.3
Chromosome Location
Chr. 6: 166929509 - 166956589 on Build GRCh38
UniGene ID
Hs.529989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003730.4 NP_003721.2 6 822
XM_017011397.1 XP_016866886.1 7 601
XM_017011398.1 XP_016866887.1 5 723
XM_017011399.1 XP_016866888.1 6 783
GenBank mRNA AJ419865.1 - 6 776
AJ419866.1 - 6 1496
AJ419867.1 - 6 730
AK001769.1 - 3 397
AK315467.1 - 6 783
BC001660.2 - 6 748 2959583
BC001819.2 - 6 748 2959583
BC036282.1 - 6 809
BC039713.1 - 6 751 5225327
BC051912.1 - 6 761 5227463
U85625.2 - 6 779

View Full Product Details