Gene Transcripts

Gene Symbol
Pcdh19
Entrez Gene ID
317183
Gene Name
protocadherin 19
Gene Aliases
RGD1565392
Chromosome Location
Chr. X: 104387346 - 104493914 on Build Rnor_6.0
UniGene ID
Rn.24970
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169129.1 NP_001162600.1 3 2502
XM_006257222.2 XP_006257284.1 4 2843
XM_006257223.2 XP_006257285.1 4 2843
XM_006257224.2 XP_006257286.1 3 2702

View Full Product Details